Skip to Content
MilliporeSigma
All Photos(1)

Documents

F8527

Sigma-Aldrich

Monoclonal Anti-CD45−FITC/CD14-PE DUAL-TAG antibody produced in mouse

clone BRA-55/UCHM-1, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

FITC conjugate
phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BRA-55/UCHM-1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 20 μL using 1 × 106 cells

isotype

IgG1/IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Application

Monoclonal Anti-CD45-FITC/CD14-PE DUAL-TAG antibody is suitable for flow cytometry at a concentration of 20μL using 1×106 cells.

Biochem/physiol Actions

CD45 is a family of single chain transmembrane glycoproteins, consisting of different isoforms, which share a common large intracellular domain. It is abundantly present in leukocyte cell surface and belongs to a novel class of enzymes, the protein tyrosine phosphatases (PTPs). It plays an essential role in thymic selection and peripheral activation of T and B cells. It is associated with various signal transduction pathways. It may also accelerate the Src family tyrosine kinases activity. an important determinant of immune tolerance.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Legal Information

DUAL-TAG is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calvin C K Yu et al.
Current directions in autoimmunity, 6, 61-88 (2002-11-01)
Mutations in a number of signaling components in mice can lead to strong autoimmune phenotypes. In some cases, these mutations likely compromise important feedback inhibitory pathways that downregulate antigen receptor signaling. For example, a deficiency of Lyn leads to a
B Tackenberg et al.
Autoimmunity, 36(2), 117-121 (2003-06-25)
In myasthenia gravis (MG), humoral and cellular immune mechanisms are involved in the autoimmune pathogenesis. In this study, we investigated the role of the CD45 molecule in MG, having recently reported an association in multiple sclerosis. CD45, a protein-tyrosine phophatase
I S Trowbridge et al.
Annual review of immunology, 12, 85-116 (1994-01-01)
CD45 is one of the most abundant leukocyte cell surface glycoproteins and is expressed exclusively upon cells of the hematopoietic system. Different isoforms of CD45 are generated by alternative splicing and are expressed in cell type-specific patterns on functional subpopulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service