Key Documents
F8395
Sigma-Aldrich
Carestream® electron microscopy film 4489
About This Item
UNSPSC Code:
41105300
NACRES:
MA.01
sheet size
3 1/4 in. × 4 in.
storage temp.
2-8°C
General description
Fine-grain, blue-sensitive emulsion on 7-mil ESTAR base, notched for emulsion identification. Tear- and abrasion-resistant, dimensionally stable. Requires shorter pump-down time and curls less than Kodak electron image film SO-163.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)