Key Documents
F8270
Sigma-Aldrich
Carestream® electron image film SO-163
About This Item
UNSPSC Code:
41105341
sheet size
6.5 cm × 9 cm
storage temp.
2-8°C
General description
Fine-grain, blue-sensitive emulsion on 7-mil ESTAR base, notched for emulsion identification. Tear- and abrasion-resistant, dimensionally stable. Greater electron exposure and development latitudes than 4489.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)