Skip to Content
MilliporeSigma
All Photos(1)

Documents

F8105

Sigma-Aldrich

MCDB 153 Medium

With HEPES and sodium bicarbonate, without ʟ-glutamine, sterile liquid, suitable for cell culture

Synonym(s):

153 Medium, Medium 153, MCDB Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

product name

MCDB 153 Medium, With trace elements, and 28mM HEPES, with sodium bicarbonate, without L-glutamine, liquid, suitable for cell culture

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

components

L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

Application

Originally developed for clonal and long term growth of human epidermal keratinocytes under serum-free conditions.

Quantity

Formulated to contain 17.6 grams of powder per liter of medium.

Reconstitution

Supplement with 1.18 g/L sodium bicarbonate.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service