Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F7376

Sigma-Aldrich

4-Fluoro-DL-tryptophan

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11FN2O2
CAS Number:
Molecular Weight:
222.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (TLC)

form

crystalline

technique(s)

NMR: suitable

color

off-white

storage temp.

−20°C

SMILES string

NC(Cc1c[nH]c2cccc(F)c12)C(O)=O

InChI

1S/C11H11FN2O2/c12-7-2-1-3-9-10(7)6(5-14-9)4-8(13)11(15)16/h1-3,5,8,14H,4,13H2,(H,15,16)

InChI key

DEBQMEYEKKWIKC-UHFFFAOYSA-N

Biochem/physiol Actions

4-Fluoro-DL-tryptophan (4-F-TRP) is used to label bacterial arginyl-tRNA synthetases for conformational analysis and to label myoglobins and hemoglobins for NMR spectra analysis.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q S Zhang et al.
Journal of protein chemistry, 18(2), 187-192 (1999-05-20)
Escherichia coli 4-fluorotryptophan-substituted arginyl-tRNA synthetase was biosynthetically prepared and purified from a tryptophan auxotroph which could overproduce this enzyme. A method was developed to separate 4-fluorotryptophan from tryptophan and to determine accurately their contents in the 4-fluorotryptophan-containing proteins. It was
P M Bronskill et al.
The Biochemical journal, 249(1), 305-308 (1988-01-01)
The tryptophan-auxotrophic Bacillus subtilis LC33 mutant strain utilizes either tryptophan or 4-fluorotryptophan for growth. Proteins therefore could be isolated from these cells in either tryptophan-containing or 4-fluorotryptophan-containing forms. Since 4-fluorotryptophan is non-fluorescent, tryptophan fluorescence would be suppressed in the 4-fluorotryptophan-containing
P Soumillion et al.
Biochemistry, 37(7), 1819-1827 (1998-03-04)
Clamp proteins confer processivity to the DNA polymerase during DNA replication. These oligomeric proteins are loaded onto DNA by clamp loader protein complexes in an ATP-dependent manner. The mechanism by which the trimeric bacteriophage T4 clamp protein (the 45 protein)
J L Hott et al.
The Biochemical journal, 264(1), 297-299 (1989-11-15)
The derivative 4-fluorotryptophan was confirmed to have negligible fluorescence at 25 degrees C and 285 nm (tryptophan/4-fluorotryptophan quantum-yield ratio greater than 100:1). However, photolysis experiments on tryptophan and 4-fluorotryptophan, in which loss of starting material was measured by reverse-phase h.p.l.c.
Christoph Giese et al.
ChemMedChem, 3(9), 1449-1456 (2008-08-30)
Nonproteinogenic amino acids that either occur naturally or are synthesized chemically are becoming important tools in modern drug discovery. In this context, fluorinated amino acids have great potential in the development of novel pharmaceuticals and drugs. To assess whether different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service