Flumequine inhibits topoisomerases, which are needed for the transcription and replication of bacterial DNA. The inhibition of the topoisomerases results in strand breakage of the bacterial chromosome, supercoiling, and resealing. Therefore, DNA replication and transcription is inhibited .
Antimicrobial agents and chemotherapy, 43(9), 2131-2137 (1999-09-03)
The occurrence of mutations in the genes coding for gyrase (gyrA and gyrB) and topoisomerase IV (parE and parC) of Salmonella typhimurium experimental mutants selected in vitro and in vivo and of 138 nalidixic acid-resistant Salmonella field isolates was investigated.
The DNA gyrases from Mycobacterium avium, Mycobacterium smegmatis and Mycobacterium fortuitum bv. peregrinum, which are species naturally resistant, moderately susceptible and susceptible to fluoroquinolones, respectively, were purified by affinity chromatography on novobiocin-Sepharose columns. The DNA gyrase inhibiting activities (IC50 values)
Toxicological sciences : an official journal of the Society of Toxicology, 69(2), 317-321 (2002-10-16)
In order to elucidate the tumor-initiating potential of flumequine (FL) in the liver, male C3H mice were given dietary administration of 4000 ppm FL throughout the study or for 2 weeks at the initiation stage, and then received 2 intraperitoneal
Journal of medical microbiology, 53(Pt 9), 895-901 (2004-08-18)
The mechanisms of resistance to quinolone and epidemiological relationships among A. salmonicida strains isolated from diseased fish in French marine farms from 1998 to 2000 were investigated. The quinolone resistance-determining regions of the gyrA and parC genes of 12 clinical
Flumequine (FLU) is used in the treatment of systemic bacterial infections in poultry, including colibacillosis, which is a common disease in turkeys. The pharmacokinetic (PK) behavior of FLU administered to 32 healthy turkeys as an oral bolus via gavage or