Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F6147

Sigma-Aldrich

Fluoranthene-3-14C

~95% (Radiochemical Purity), methanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

~95% (Radiochemical Purity)

form

methanol solution

extent of labeling

40-60 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

c1ccc-2c(c1)-c3cccc4[14cH]ccc-2c34

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service