Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F5146

Sigma-Aldrich

Fast Red RC

tablet, 10 mg substrate per tablet

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

tablet

solubility

deionized water: 1 tablet/10 mL, clear, pale yellow

storage temp.

room temp

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1ccc(Cl)cc1[N+]#N.COc2ccc(Cl)cc2[N+]#N

InChI

1S/2C7H6ClN2O.4ClH.Zn/c2*1-11-7-3-2-5(8)4-6(7)10-9;;;;;/h2*2-4H,1H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

YHUGASVQWCSKGW-UHFFFAOYSA-J

Specificity

Fast Red RC is used in conjunction with the alkaline phosphatase substrate Naphthol AS-TR Phosphate (Product No. N8518) in immunoblotting and immunohistological staining procedures. This substrate system provides an insoluble end product that is red in color and can be observed visually. Fast Red RC Tablets may also be used with other naphthol phosphate derivatives and in other applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pluzek, K.-J., Ramlau, J. et al.
Handbook of Immunoblotting of Proteins, 1, 177-177 (1988)
Larsson, L.-I.
Immunocytochemistry: Theory and Practice, 95-95 (1988)
Ana M Martín-Hernández et al.
Journal of virology, 82(8), 4064-4071 (2008-02-15)
RNA silencing is a host defense mechanism that limits the accumulation and spread of viruses in infected plants. Correspondingly, plant viruses encode suppressors of silencing. In the positive-strand RNA virus Tobacco rattle virus (TRV), the suppressor of silencing is a
Florian R Fritzsche et al.
BMC cancer, 8, 381-381 (2008-12-23)
Enhanced activity of histone deacetylases (HDAC) is associated with more aggressive tumour behaviour and tumour progression in various solid tumours. The over-expression of these proteins and their known functions in malignant neoplasms has led to the development of HDAC inhibitors
Jutta Schade et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 56(2), 147-155 (2007-10-31)
The expression of dipeptidyl peptidase 4 (DP4, CD26) affects T-cell recruitment to lungs in an experimental rat asthma model. Furthermore, the gene of the structural homologous DP10 represents a susceptibility locus for asthma in humans, and the functional homologous DP8/9

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service