Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F4521

Sigma-Aldrich

Fluorescein di-(β-D-glucopyranoside)

≥90%

Synonym(s):

FDGlu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32O15
CAS Number:
Molecular Weight:
656.59
Beilstein:
7455945
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥90%

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3c(Oc4cc(O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)ccc4C36OC(=O)c7ccccc67)c2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C32H32O15/c33-11-21-23(35)25(37)27(39)30(45-21)42-13-5-7-17-19(9-13)44-20-10-14(43-31-28(40)26(38)24(36)22(12-34)46-31)6-8-18(20)32(17)16-4-2-1-3-15(16)29(41)47-32/h1-10,21-28,30-31,33-40H,11-12H2/t21-,22-,23-,24-,25+,26+,27-,28-,30-,31-/m1/s1

InChI key

ZTOBILYWTYHOJB-LWXMCBIJSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service