Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F3679

Sigma-Aldrich

Anti-Frizzled-7 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FZD7

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4 μg/mL using human kidney

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FZD7(8324)
mouse ... Fzd7(14369)

General description

Frizzled homolog protein (FZD) is a seven-pass transmembrane type receptor expressed in various normal tissues such as skeletal muscle, kidney, pancreas, cerebellum and cerebral cortex. In human, ten members (FZD1-FZD10) of Frizzled homolog protein have been identified. FZD7 is expressed throughout normal gastrointestinal tract, from esophagus to rectum. It is highly expressed in the gastric cancer cell line MKN7 and relatively lowers in esophageal cancer cell lines.

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human frizzled-7, conjugated to KLH. The immunizing peptide has 100% homology with the mouse gene.

Application

Anti-Frizzled-7 antibody is suitable for immunostaining of liver tissues to localise FZD7 in T and pT tissues. It is also suitable for immunohistochemistry (formalin-fixed, paraffin-embedded sections) at a concentration of 4μg/mL using human kidney.

Biochem/physiol Actions

FZD7 shows 98.8% nucleotide identity with FzE3 and encodes for WNT receptors with nine amino-acid substitutions. In human primary gastric cancer, high level of FZD7 has been found. Mechanistically, over-expression of Frizzled-7 accelerates the WNT-β-catenin-TCF pathway. Through the activation of the WNT-β-catenin-TCF pathway, FZD7, might play key roles in carcinogenesis.

Physical form

Solution in phosphate buffered saline containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Bengochea et al.
British journal of cancer, 99(1), 143-150 (2008-06-26)
Dysregulation of growth factors and their receptors is central to human hepatocellular carcinoma (HCC). We previously demonstrated that the Frizzled-7 membrane receptor mediating the Wnt signalling can activate the beta-catenin pathway and promotes malignancy in human hepatitis B virus-related HCCs.
H Kirikoshi et al.
International journal of oncology, 19(1), 111-115 (2001-06-16)
Human Frizzled-7 (FZD7) and human FzE3, showing 98.8% nucleotide identity, encode almost identical WNT receptors with nine amino-acid substitutions. FzE3 is claimed to be expressed specifically in esophageal cancer. We determined the structure of the FZD7 gene and the FZD7
Koji Ueno et al.
International journal of cancer, 132(8), 1731-1740 (2012-07-27)
Wnt signaling pathways play important roles in tumorigenesis and are initiated by binding of Wnt to various receptors including frizzleds (FZDs). FZDs are one of several families of receptors comprised of FZD/LRP/ROR2/RYK in the Wnt signaling pathway. Expression of some
H Kirikoshi et al.
International journal of oncology, 19(4), 767-771 (2001-09-20)
Frizzled (FZD) genes encode seven-transmembrane type WNT receptors, which are implicated in carcinogenesis and embryogenesis. We have previously cloned and characterized FZD1, FZD2, FZD3, FZD4, FZD5, FZD6, FZD7, FZD8, and FZD10. Here, we investigated expression profiles of all members of
Sze Chuen Cesar Wong et al.
PloS one, 8(11), e79481-e79481 (2013-11-21)
Frizzled homolog 3 receptor was up-regulated in several gastrointestinal cancers such as esophageal and gastric cancers. Moreover, frizzled homolog 3 has recently reported to be expressed in colorectal adenoma specimens. In the present study, we investigated the clinical significance of

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service