Skip to Content
MilliporeSigma
All Photos(1)

Documents

F3641

Sigma-Aldrich

Firefly Lantern Extract

powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.25

form

powder

storage temp.

−20°C

Packaging

Package size based on weight of dried lanterns extracted

Physical form

Glycine buffered, with added luciferin

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Varma et al.
Ophthalmologica. Journal international d'ophtalmologie. International journal of ophthalmology. Zeitschrift fur Augenheilkunde, 219(5), 309-315 (2005-08-27)
Cataract is one of the most significant vision-impairing complications of diabetes. The present study examined the feasibility of inhibiting cataract formation by treatment with pyruvate, a metabolite known to effectively scavenge reactive species of oxygen and inhibit protein glycation, both
A R Ribeiro et al.
Journal of bioluminescence and chemiluminescence, 13(6), 371-378 (1999-02-02)
The bioluminescent reaction catalysed by firefly luciferase has become widely established as an outstanding analytical system for assay of ATP. When used in solution, luciferase is unstable and cannot be re-used, a problem that can be partially circumvented by immobilizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service