Skip to Content
MilliporeSigma
All Photos(1)

Documents

F3177

Sigma-Aldrich

Fluorescein-α-D-galactosamine polyacrylamide

powder

Synonym(s):

α-D-GalNAc-PAA-FI

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

powder

Quality Level

mol wt

~30 kDa

color

yellow

storage temp.

2-8°C

Application

Fluorescein-α-D-galactosamine polyacrylamide may be used in the development of cross-linkable hydrogels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lihui Weng et al.
Journal of vascular and interventional radiology : JVIR, 22(10), 1464-1470 (2011-08-06)
To develop and evaluate a bioresorbable spherical material for embolization. New bioresorbable hydrogel microspheres were prepared from carboxymethyl cellulose and chitosan by using an inverse emulsion method. Size distribution of the microspheres was determined with a microscope, and the colorability
Lihui Weng et al.
Biomaterials, 29(29), 3905-3913 (2008-07-22)
A series of in situ gelable hydrogels were prepared from oxidized dextran (Odex) and N-carboxyethyl chitosan (CEC) without any extraneous crosslinking agent. The gelation readily took place at physiological pH and body temperature. The gelation process was monitored rheologically, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service