Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F2922

Sigma-Aldrich

Monoclonal Anti-FLAG® BioM5−Biotin antibody produced in mouse

clone M5, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M5, monoclonal

form

buffered aqueous solution

technique(s)

western blot (chemiluminescent): 2 μg/mL

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

General description

Anti-FLAG® BioM5 monoclonal antibody is a purified murine IgG1 monoclonal antibody that is covalently attached by hydrazide linkage. It can be detected by avidin or streptadivin conjugates.

Application

Anti-FLAG® BioM5 monoclonal antibody is useful for Western blotting, microscopy applications and formation of avidin-biotin complexes (ABC) in mammalian and Drosophila cells. Anti-FLAG® BioM5 antibody in combination with an avidin or streptavidin conjugate is the preferred anti-FLAG® antibody for detection of FLAG fusion proteins expressed in mouse cells.
The product binds the FLAG peptide only when it is located at the amino terminus preceded by a methionine. Binding is not Ca2+-dependent. It is useful for detecting cytoplasmically expressed Met-FLAG® fusion proteins in mammalian crude cell extracts, but not recommended for fusion proteins expressed in E. coli.
It can be used for immunodetection methods using avidin- or streptavidin-conjugated reporter enzymes such as streptavidin-peroxidase. Primary antibody conjugates are preferred when using murine cells as the recombinant protein host.

Browse additional application references in our FLAG® Literature portal.
Binds the FLAG peptide only when it is located at the amino terminus preceded by a methionine. Binding is not Ca2+-dependent. Useful for detecting cytoplasmically expressed Met-FLAG fusion proteins in mammalian crude cell extracts, but not recommended for fusion proteins expressed in E. coli.

Features and Benefits

ANTI-FLAG M5 − Biotin conjugate has a high affinity for N-terminal Met-FLAG fusion proteins.

Physical form

Solution in 10 mM sodium phosphate, pH 7.4, containing 150 mM NaCl and 0.02% sodium azide

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinna Zhang et al.
The EMBO journal, 30(11), 2177-2189 (2011-04-28)
Tumour suppressor p53 levels in the cell are tightly regulated by controlled degradation through ubiquitin ligases including Mdm2, COP1, Pirh2, and ARF-BP1. The ubiquitination process is reversible via deubiquitinating enzymes, such as ubiquitin-specific peptidases (USPs). In this study, we identified
Jin-Ho Koh et al.
Cell metabolism, 25(5), 1176-1185 (2017-05-04)
The objective of this study was to evaluate the specific mechanism(s) by which PPARβ regulates mitochondrial content in skeletal muscle. We discovered that PPARβ increases PGC-1α by protecting it from degradation by binding to PGC-1α and limiting ubiquitination. PPARβ also
Peipei Wang et al.
The Journal of biological chemistry, 299(8), 105055-105055 (2023-07-17)
Post-translational modifications including protein ubiquitination regulate a plethora of cellular processes in distinct manners. RNA N6-methyladenosine is the most abundant post-transcriptional modification on mammalian mRNAs and plays important roles in various physiological and pathological conditions including hematologic malignancies. We previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service