Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F2016

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

indirect immunofluorescence: 1:160 using mouse splenocytes

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binds all goat Igs.
IgG (immunoglobulin G) antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids.

Application

Anti-Goat IgG (whole molecule)-FITC antibody produced in rabbit has been used in immunofluorescence to study the translocation of CXCR4 in human hepatoma cell lines. It has also been used in immunocytochemistry.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of FITC to Anti-Goat IgG (whole molecule) antibody allows for the visualization of protein by fluorescent microscopy.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Song et al.
Stem cells and development, 14(6), 712-721 (2006-01-26)
Much of the knowledge regarding the regulatory pathways for adult stem cell self-renewal and differentiation has been obtained from the results of in vitro cultures. However, it is unclear if adult stem cells are controlled in the same way under
Jeremiah Morrissey et al.
Journal of the American Society of Nephrology : JASN, 11(9), 1681-1690 (2000-08-31)
Analysis of gene expression in a mouse model of unilateral ureteral obstruction (UUO) revealed significant induction of CD14 mRNA in kidneys with obstructed ureters. Immunocytochemical analysis indicated that CD14 was upregulated in tubular epithelial cells and this upregulation was not
Ramadan F Abdelaziz et al.
International journal of molecular sciences, 24(23) (2023-12-09)
Cancer is one of the main causes of death globally. Radiotherapy/Radiation therapy (RT) is one of the most common and effective cancer treatments. RT utilizes high-energy radiation to damage the DNA of cancer cells, leading to their death or impairing
Marta de Almeida Santiago et al.
PloS one, 11(12), e0167669-e0167669 (2016-12-10)
The use of antibodies in immunodiagnostic kits generally implies the conjugation of these proteins with other molecules such as chromophores or fluorochromes. The development of more sensitive quality control procedures than spectrophotometry is essential to assure the use of better
C C Schimanski et al.
British journal of cancer, 95(2), 210-217 (2006-07-05)
In different tumour entities, expression of the chemokine receptor 4 (CXCR4) has been linked to tumour dissemination and poor prognosis. Therefore, we evaluated, if the expression of CXCR4 exerts similar effects in human hepatocellular carcinoma (HCC). Expression analysis and functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service