Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

F1879

Sigma-Aldrich

Formaldehyde Dehydrogenase from Pseudomonas sp.

lyophilized powder, 1.0-6.0 units/mg solid

Synonym(s):

FDH, Formaldehyde:NAD+ oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

1.0-6.0 units/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde Dehydrogenase (FDH) belongs to the medium-chain alcohol dehydrogenase family. It is encoded by the fdhA and fdhB gene in Pseudomonas aeruginosa. The FDH sequences of P. aeruginosa and P. putida are conserved and show homology. FDH exists as dimer and tetramer with zinc in their catalytic active site. The monomer corresponds to 350-400 amino-acid residues.

Application

Formaldehyde Dehydrogenase from Pseudomonas sp. has been used:

  • For standard curve generation for the demethylation-FDH assay of lysine specific demethylase 1 (LSD1) and in formaldehyde dehydrogenase (FDH) assay.
  • As a biosensor for the presence of formaldehyde in pharmaceuticals, waste water, vaccines and industrial products.
  • In coupled pectin methyl esterase (PME) enzyme assay.

Biochem/physiol Actions

Formaldehyde dehydrogenase (FDH) is an enzyme that catalyzes the oxidation of formaldehyde to formate with NAD+ as an electron acceptor.Formaldehyde dehydrogenases are involved in the biological detoxification of formaldehyde, a toxic product in the metabolism of compounds like histidine, vanillate etc. They detoxify by converting formaldehyde to carbon-di-oxide. FDH plays a crucial role in regulating protein S-nitrosation in mammalian cells by decomposing S-nitrosoglutathione. They are also involved in the regulation of nitrosothiol homeostasis by catalyzing the reduction of endogenous nitrosylating agent S-nitrosoglutathione (GSNO).

Unit Definition

One unit will oxidize 1.0 μmole of formaldehyde to formic acid per min at pH 7.5 at 37 °C.

Physical form

Lyophilized powder containing ~70% stabilizers as Mg2+, Ca2+, bovine serum albumin, glycine, and lysine

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of a formaldehyde dehydrogenase-glutathione pathway in protein S-nitrosation in mammalian cells
Haqqani A S, et al.
Nitric Oxide, 9(3), 172-181 (2003)
Structure of formaldehyde dehydrogenase from Pseudomonas aeruginosa: the binary complex with the cofactor NAD+
Liao Y, et al.
Acta Crystallographica. Section F, Structural Biology Communications, 69(9), 967-972 (2013)
Expression, purification, and characterization of formaldehyde dehydrogenase from Pseudomonas aeruginosa
Zhang W, et al.
Protein Expression and Purification, 92(2), 208-213 (2013)
Ivan V Rosado et al.
Nature structural & molecular biology, 18(12), 1432-1434 (2011-11-15)
Metabolism is predicted to generate formaldehyde, a toxic, simple, reactive aldehyde that can damage DNA. Here we report a synthetic lethal interaction in avian cells between ADH5, encoding the main formaldehyde-detoxifying enzyme, and the Fanconi anemia (FA) DNA-repair pathway. These
Redundancy of enzymes for formaldehyde detoxification in Pseudomonas putida
Roca A, et al.
Journal of Bacteriology, 191(10), 3367-3374 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service