Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1642

Sigma-Aldrich

Flurandrenolide

Synonym(s):

6α-Fluoro-11β,16α,17,21-tetrahydroxypregn-4-ene-3,20-dione 16,17-acetonide, Fludroxycortide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H33FO6
CAS Number:
Molecular Weight:
436.51
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CC1(C)O[C@@H]2CC3C4C[C@H](F)C5=CC(=O)CCC5(C)C4C(O)CC3(C)[C@@]2(O1)C(=O)CO

InChI

1S/C24H33FO6/c1-21(2)30-19-9-14-13-8-16(25)15-7-12(27)5-6-22(15,3)20(13)17(28)10-23(14,4)24(19,31-21)18(29)11-26/h7,13-14,16-17,19-20,26,28H,5-6,8-11H2,1-4H3/t13?,14?,16-,17?,19+,20?,22?,23?,24+/m0/s1

InChI key

POPFMWWJOGLOIF-CCRGDLLISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E L Jones et al.
Cutis, 21(6), 881-885 (1978-06-01)
New methods are now available to help select vehicles for topical corticosteroids. In the development of a new cream vehicle for flurandrenolide, the goal was a cream base with optimal solubility for release of the drug, minimal irritation potential, and
H Nakamura et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 76(7), 595-607 (1980-10-01)
Fludroxycortide tape is a thin plastic tape which contains a synthetic glucocorticoid, fludroxycortide of 4 microgram/cm2. fludroxycortide tape with a topical application of 1 cm2 inhibited significantly contact hypersensitivities to oxazolone and picryl chloride in the ear skin of mice
B M Hausen et al.
Dermatosen in Beruf und Umwelt. Occupation and environment, 33(1), 27-28 (1985-01-01)
A 66-year-old female developed increasing eczematous reactions after treatment of her leg ulcer with a corticoid ointment. When patch tested with the constituents she reacted positive to fludroxycortide (flurandrenolone) and cetyl alcohol. Contact allergy to this fluorinated corticosteroid has not
T Lucke et al.
Clinical and experimental dermatology, 24(4), 266-269 (1999-08-24)
The formation of milia is well recognized in both bullous and inflammatory dermatoses. There are several reports of milia developing in a rare variant of lichen planus pilaris known as lichen planus follicularis tumidus (LPFT), but the association of milia
Setaluri V et al.
Journal of cutaneous medicine and surgery, 4(4), 196-198 (2001-03-07)
Flurandrenolide tape is a valuable agent in the treatment of lichen simplex chronicus and in psoriasis. The corticosteroid is helpful for reducing the symptoms and induration of the lichen simplex chronicus lesion, and occlusion of the lesion with the tape

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service