F1375
Fast Red ITR Salt
practical grade
Synonym(s):
5-Diethylaminosulfonyl-2-methoxybenzenediazonium chloride hemi(zinc chloride) salt
About This Item
Recommended Products
grade
practical grade
storage temp.
−20°C
SMILES string
CCN(CC)S(=O)(=O)c1ccc(OC)c(c1)[N]#N
InChI
1S/C11H16N3O3S/c1-4-14(5-2)18(15,16)9-6-7-11(17-3)10(8-9)13-12/h6-8H,4-5H2,1-3H3
InChI key
DLMVAZUSXBWNAJ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service