Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

F0891

Sigma-Aldrich

Flumethasone pivalate

Synonym(s):

6α,9-Difluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 21-(2,2-dimethylpropionate), Flumetasone 21-pivalate, NSC 107680

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36F2O6
CAS Number:
Molecular Weight:
494.57
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:
NACRES:
NA.85

form

solid

optical activity

[α]25/D 71.0 to 82.0°, c = 1 in dioxane

solubility

dioxane: 4.0 mL, clear, colorless to very faintly yellow (50 mg + 4.0 mL dioxane)

Mode of action

cell membrane | interferes

SMILES string

[H][C@@]12C[C@@H](C)[C@](O)(C(=O)COC(=O)C(C)(C)C)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])C[C@H](F)C4=CC(=O)C=C[C@]34C

InChI

1S/C27H36F2O6/c1-14-9-16-17-11-19(28)18-10-15(30)7-8-24(18,5)26(17,29)20(31)12-25(16,6)27(14,34)21(32)13-35-22(33)23(2,3)4/h7-8,10,14,16-17,19-20,31,34H,9,11-13H2,1-6H3/t14-,16+,17+,19+,20+,24+,25+,26+,27+/m1/s1

InChI key

JWRMHDSINXPDHB-OJAGFMMFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Flumethasone pivalate is a topical difluorinated corticosteroid ester with anti-inflammatory, antipruritic and vasoconstrictive properties. A prompt decrease in inflammation, exudation and itching is experienced after application. It is used to study adrenocortical suppression, plasma transcortin binding, and cutaneous atrophy and telangiectasia.

Biochem/physiol Actions

Flumethasone pivalate is a glucocorticoid receptor agonist used in studies on plasma transcortin binding. It binds to the nucleus causing a variety of genetic activations and repressions. The anti-inflammatory action of flumethasone is thought to involve lipocortins and phospholipase A2 inhibitory proteins, which results in the inhibition of arachidonic acid and the control of prostaglandin and leukotriene biosynthesis. Flumethasone suppresses the immune system due to a decrease in the function of the lymphatic system, a reduction in immunoglobulin and complement concentrations, the precipitation of lymphocytopenia, and interference with antigen-antibody binding.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Hojyo
The Journal of international medical research, 15(5), 255-263 (1987-09-01)
The combination creams, betamethasone dipropionate/clotrimazole/gentamicin sulphate and flumethasone pivalate/clioquinol, were compared in patients with corticosteroid responsive dermatoses and/or cutaneous fungal and/or bacterial infections. Medication was applied to affected areas twice daily for 28 days. Of 67 patients enrolled, 31 treated
Corticosteroid-Induced Cutaneous Atrophy and Telangiectasia: Experimental Production Associated With Weight Loss in Rats
. Graham Smith, Jr, Richard F. Wehr, et al.
Archives of Dermatological Research, 112, 1115-1117 (1976)
Adrenocortical Suppression With Topical Flumethasone
Richard D. Carr and Ralph G. Wieland
Archives of Dermatological Research, 96, 269-272 (1967)
[Release of flumethasone pivalate from oleogel and W/O emulsion bases in vitro].
W Horsch et al.
Die Pharmazie, 37(11), 807-808 (1982-11-01)
Analysis of flumethasone pivalate formulations by high-performance liquid chromatography.
B A Lodge et al.
Journal of chromatography, 301(2), 477-480 (1984-10-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service