Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F0877

Sigma-Aldrich

D-Fructose 1-phosphate barium salt

≥97%, crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11O9PBa
CAS Number:
Molecular Weight:
395.45
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥97%

form

crystalline

storage temp.

−20°C

SMILES string

[Ba].OCC(O)C(O)C(O)C(=O)COP(O)(O)=O

Application

D-Fructose 1-phosphate is an effector of the metabolic regulator Cra of Pseudomonas putida and inhibitor of translation of SugR, a pleiotropic transcriptional repressor, in Corynebacterium glutamicum.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M Ramseier
Research in microbiology, 147(6-7), 489-493 (1996-07-01)
The catabolite repressor-activator (Cra) protein controls the direction of carbon flux through metabolic pathways in enteric bacteria. Cra binds to the control regions of target genes and exerts a negative effect on the expression of genes encoding glycolytic and Entner-Doudoroff
Christiane Dietrich et al.
Microbiology (Reading, England), 155(Pt 4), 1360-1375 (2009-04-01)
Corynebacterium glutamicum is a biotin-auxotrophic bacterium and some strains efficiently produce glutamic acid under biotin-limiting conditions. In an effort to understand C. glutamicum metabolism under biotin limitation, growth of the type strain ATCC 13032 was investigated in batch cultures and
Max Chavarría et al.
The Journal of biological chemistry, 286(11), 9351-9359 (2011-01-18)
The catabolite repressor/activator (Cra) protein is a global sensor and regulator of carbon fluxes through the central metabolic pathways of gram-negative bacteria. To examine the nature of the effector (or effectors) that signal such fluxes to the protein of Pseudomonas
Haruhiko Teramoto et al.
Journal of biotechnology, 154(2-3), 114-125 (2011-02-01)
Corynebacterium glutamicum, a high-GC Gram-positive soil bacterium, has been used in development of bioprocesses for production of various compounds such as amino acids, organic acids, and alcohols. Recently, several transcriptional regulators, each of which is involved in multiple carbon metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service