Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EXTRA3

Sigma-Aldrich

ExtrAvidin® Peroxidase Staining Kit for rabbit antibodies antibody produced in mouse

Synonym(s):

ExtrAvidin® Peroxidase Staining Kits

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

antibody product type

secondary antibodies

usage

sufficient for 2,000 tests ELISA, Dot blot
sufficient for 200 tests Immunohistology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

These kits comprise universal reagents for use with primary antibodies in immunohistology, ELISA, and immunoblotting.
ExtrAvidin® is a unique form of avidin, available only from Sigma, that combines the high specificity and affinity of avidin for biotin with low non-specific binding at physiological pH. ExtrAvidin® peroxidase exhibits high sensitivity with low background.

Features and Benefits

  • Use in immunohistology, ELISA, and immunoblotting assays.
  • Affinity Isolated Antibodies have been adsorbed with human IgG and IgM to minimize cross-reactivity.
  • Biotinylated antibodies contain a spacer which improves accessibility for the ExtrAvidin® conjugates.

Components

• 3 mL ExtrAvidin®-Peroxidase
• 3 mL Biotinylated Secondary Antibodies
• Complete Instructions and Assay Protocols

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Wang et al.
Prostaglandins & other lipid mediators, 143, 106343-106343 (2019-06-14)
Prostaglandins (PGs) play a pivotal role in uterine reproductive process including maternal recognition of pregnancy, cell proliferation, and myometrium contractions in mammals. In this study, we investigated the immunolocalizations and expression levels of Prostaglandin E2 synthases cyclo-oxygenase (COX)-1 and COX-2
Vijayalaxmi et al.
Neuropeptides, 81, 101999-101999 (2019-12-18)
Enkephalins are the pentapeptides involved in pain relief and neuroendocrine responses with high affinity for delta opioid receptors in vertebrates. In the present investigation, we studied the distribution of leucine-enkephalin-immunoreactive (L-ENK-ir) neurons in the brain of the cichlid fish Oreochromis
Zhu Chen et al.
Prostaglandins & other lipid mediators, 148, 106412-106412 (2020-01-14)
The prostate gland is a male accessory reproductive gland, whose vitality and function are under tight regulation of different hormones. Prostaglandins E2 (PGE2) is one of the major products generated by the actions of cyclooxygenases (COX) and prostaglandin E synthase
Ana Navarro et al.
Frontiers in aging neuroscience, 10, 254-254 (2018-09-07)
Apolipoprotein D (Apo D) is a key molecule in the lipid transport during homeostasis and repair processes in normal and pathological conditions of the nervous system with a putative neuroprotective effect. In the last decades, huge experimental efforts have been
Ritu Kulshrestha et al.
Iranian journal of basic medical sciences, 23(3), 396-405 (2020-05-23)
The role of N-acetylcysteine (NAC) as an anti-oxidant in attenuating bleomycin-induced pulmonary fibrosis has been reported. However, its effect on parenchymal remodeling via regulating the protease-antiprotease balance is not fully defined. Therefore, the present study was designed to explore the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service