EPS44I211005
Eppendorf® Innova® S44i Shaker
refrigerated, photosythetic LED light bank, 1" orbit, AC/DC input 120 V
Select a Size
$57,960.00
Select a Size
About This Item
$57,960.00
Recommended Products
feature
photosythetic LED light bank, 1" orbit
refrigerated
manufacturer/tradename
Eppendorf® S44I211005
Related Categories
General description
Product Information
The Innova S44i is a large capacity, temperature controlled (refrigerated) stackable shaker allowing the use of high-volume flasks in large numbers in a compact footprint. The iconic slide up door provides easy access to your samples. The Eppendorf X-Drive allows for shaking high loads at high speeds – even when stacked. When there is an instance of an unbalanced load, the X-Drive′s counterbalance adjustment allows for smooth shaking. The optional photosynthetic LED lightbank provides the ability to evenly light the entire platform across a wide intensity range – and it′s programmable. The VisioNize touch interface introduces onboard monitoring, user management and the ability to export data via USB; or network integration via ethernet. A comprehensive service offering including IQ/OQ is also available.
Maximum capacity
- The Innova S44i has optimized chamber, platform, and features to maximize your options and capacity.
- Up to 136 % more capacity than the previous Innova 44 with a smaller footprint
- Up to 102 % higher flask capacity than competitor
- Double- or triple-stack the Innova S44i to increase capacity without sacrificing more valuable lab space
Eppendorf X-Drive
- The Eppendorf X-Drive is precisely engineered with five-eccentric shafts to balance even the heaviest load while providing consistent and smooth shaking.
- The intelligent counterbalance technology of the Eppendorf X-Drive ensures vibration-free shaking even with uneven loads. This advanced technology senses imbalance and decelerates to a safe speed to protect you and your cultures.
Advanced control
The new touchscreen user interface of the Innova S44i offers more than just an easy operation. This multi-touch capacitive display makes all the information you need
easy to monitor and track.
- Information is just one touch away
- Intuitive operating, distinct design
- Manage multiple users with different levels of authorization
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)