Key Documents
EPS44I041006
Eppendorf® Mandatory stacking kit; Innova S44i, to allow installation
About This Item
UNSPSC Code:
41103800
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® S44I041006
General description
The Eppendorf Stacking Kit is designed for the triple stacking of the Innova S44i shakers, enabling users to maximize lab space and increase productivity. This stacking kit provides a secure and stable platform for placing a second shaker on top of the first S44i unit. By utilizing this kit, researchers can optimize their work area without sacrificing performance. Easy to install and compatible with the S44i model, this stacking kit is an ideal solution for laboratories aiming to streamline their operations efficiently.
Other Notes
Maximum agitation speed of Innova S44i reduced to 300 rpm when stacked on one Innova 44/44R and 250 rpm when triple stacked
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)