Key Documents
EPM1289-0001
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
holds 48 x 1.5 mL Eppendorf tubes (8-11 mm diam.)
Select a Size
1 EA
$560.00
$560.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$560.00
About This Item
UNSPSC Code:
41122804
NACRES:
NB.73
$560.00
Please contact Customer Service for Availability
feature
holds 48 x 1.5 mL Eppendorf tubes (8-11 mm diam.)
manufacturer/tradename
Eppendorf® M1289-0001 (Global)
size
300x80x129 mm
General description
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)