Key Documents
EPM1190-9921
Platform Clamps, for Eppendorf® Shakers
for Eppendorf® New Brunswick Innova, I-Series and Excella shakers, starter kit
Select a Size
1 EA
$1,000.00
$1,000.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$1,000.00
About This Item
UNSPSC Code:
41122413
$1,000.00
Please contact Customer Service for Availability
feature
for Eppendorf® New Brunswick Innova, I-Series and Excella shakers
includes 4x 250 mL, 4x 500 mL, 2x 1 L, and 2x 2 L flask clamps
starter kit
manufacturer/tradename
Eppendorf® M1190-9921 (NA)
Related Categories
General description
Flask clamps are of a single piece stainless-steel construction (not spot welded or riveted) for added strength and safety. Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds. Flask clamps come complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)