Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPI023

Sigma-Aldrich

Adenosine Deaminase Activity Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

Adenosine Deaminase (ADA) (E.C. 3.5.4.4.) is an enzyme that catalyzes the conversion of adenosine and 2′-deoxyadenosine to inosine and 2′-deoxyinosine. Adenosine Deaminase is widely distributed in various tissues and cells. There are two isoforms, ADA1 and ADA2. ADA1 is widely expressed in most cells in the body, particularly in lymphocytes and macrophages. It is present in the cytosol, nucleus and found associated with dipeptidyl peptidase-4 on the cell membrane. ADA2 was first found in the spleen but is predominantly found in the plasma and serum. Increased serum ADA levels are found in certain infectious diseases such as tuberculosis and various liver diseases such as acute hepatitis, alcoholic hepatic fibrosis, chronic active hepatitis to name a few. Adenosine Deaminase is also a marker for T-lymphocyte proliferation.

Suitability

Suitable for the detection of Adenosine Deaminase (ADA) activity in purified recombinant protein, nuclear extract and cell and tissue lysate

Principle

Inosine formed from the breakdown of adenosine is converted to uric acid with ADA Convertor and ADA Developer. The uric acid is measured at A293 nm. The kit measures total activity of adenosine deaminase with limit of quantification of 1 mU of recombinant Adenosine Deaminase.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service