Key Documents
EPAG7-TT16
Test Tube Rack, for Eppendorf® New Brunswick Innova® 3100 Water Bath Shakers
holds 36 x 16 mm tubes
Select a Size
1 EA
$621.00
$621.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$621.00
About This Item
UNSPSC Code:
41122804
NACRES:
NB.73
$621.00
Please contact Customer Service for Availability
material
stainless steel
feature
holds 36 x 16 mm tubes
manufacturer/tradename
Eppendorf® AG7-TT16 (North America)
General description
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)