Key Documents
EP6731070166
Eppendorf® Door Lock For CellXpert CO2 Incubator
retrofittable
Synonym(s):
Eppendorf® Door Lock For CellXpert CO2 Incubator, Incubators
Select a Size
1 EA
$396.00
$396.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$396.00
About This Item
UNSPSC Code:
41121800
$396.00
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 6731070166
Related Categories
General description
Door lock to fit CellXpert C170i CO2 incubators. Fully retrofittable by end user and requires no service engineer.
Features and Benefits
  • Retrofittable to all CellXpert C170i produced after January 2022 (Serial number > 6731LH322982)
  • No Eppendorf Service required for installation
  • No control by user management system or open/close status tracking
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)