Skip to Content
MilliporeSigma
All Photos(1)

Documents

EP6338000012

Eppendorf® Mastercycler® Nexus X2 Thermal Cyclers

Nexus GX2e, satellite unit, aluminum block, AC/DC input 230 V, 50/60 Hz (EU)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NC.21

feature

CSA and CE compliant
Nexus GX2e
aluminum block
holds 64/32?×?0.2?mL PCR tubes or 24/10?×?0.5 mL PCR tubes
satellite unit
unit can only be controlled

manufacturer/tradename

Eppendorf® 6338000012 (EU)

parameter

1-12 °C (gradient range)
30-99 °C temp. range (gradient)
37-110 °C temp. range (Lid)
4-99 °C temp. range (control)

technique(s)

PCR: suitable

W × D × H

25 cm × 41.2 cm × 32.1 cm

weight

11 kg

Looking for similar products? Visit Product Comparison Guide

General description

The Mastercycler nexus X2 gives you the ability to run two totally independent protocols at the same time. Smaller assays fit nicely on the 32-well-block – larger assays can exceed 48 samples and run on the 64-well-block. The larger block is available with a gradient function. As every member of the Mastercycler nexus family, the Mastercycler nexus X2 can be combined with all other family members in a network of up to 3 units. In combination with Eppendorf′s PCR Tubes, PCR Strips or divisible plates, the Mastercycler nexus X2 will give you consistent and publishable results every day!
Certified Quality
All Eppendorf cyclers follow these quality guidelines:
  • Individual, documented quality control certificates
  • Calibration accordingly to national and international standards: NIST (USA), DKD/PTB (Germany), UKAS/NPL (UK)
  • UL/cUL listed

Application

Standard PCR, Fast PCR, PCR optimization

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service