Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5948000911

Eppendorf® Centrifuge 5920 R G

new

with rotor S-4xUniversal-large, bucket and tube/plate adapters, AC/DC input 230 V (GB plug)

Synonym(s):

Centrifuge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701

feature

refrigerated
to hold 68 × 15 mL OR 36 × 50 mL conical tubes OR 120 × 13 mm OR
104 × 16 mm blood collection tubes OR 20 × MTP plates OR 4 × DWP plates

with rotor S-4xUniversal-large, bucket and tube/plate adapters

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5948000911

parameter

13,700 rpm speed
−11 °-40 ° °C temp. range

Looking for similar products? Visit Product Comparison Guide

General description

Centrifuge 5920 R features an extraordinary high capacity and performance.
Its high capacity of up to 4 × 1,000 mL bottles, 28 × PCR plates or 196 × 13 mm round-bottom tubes makes it the ideal instrument for high-throughput applications.
Eppendorf developed the Dynamic Compressor Control (DCC). Unlike other compressors which switch back and forth between 0 % and 100 % performance output, DCC allows for a constant and smooth temperature regulation. This results in a precise cooling performance and the set temperature is kept stable at all time.
In addition to the standard FastTemp pre-cooling program Centrifuge 5920 R features a software option called FastTemp pro. This program allows for automated precooling based on pre-programmable time and date.
FastTemp pro can be set to a specific date or as a repetitive event for several days every week. Simply turn the centrifuge into standby mode when you leave your lab and let FastTemp pro take care of pre-cooling in the morning. This not only makes operation super easy, it also saves you more than 90 % of energy over night.
Centrifuge 5920 R comes with an automatic rotor recognition. This feature detects a newly inserted rotor and automatically limits the centrifuge speed to the rotor′s maximum permissible value.

When handling substances that pose a health hazard it is highly recommended to use aerosol-tight fixed-angle rotors with Eppendorf QuickLock lids or swing-bucket rotors in combination with Eppendorf QuickLock caps.

> Automatic rotor recognition and imbalance detection for maximum operational safety
> Sturdy motor driven lid locks to ensure secure operation
> Not one but two gas springs to support the lid
> Aerosol-tight rotors and buckets to avoid contamination

For ergonomic lid operation Eppendorf designed the soft-touch lid closure system. There is no need to exert unnecessary force when closing the centrifuge lid. In combination with a low opening height you can close the centrifuge with ease.
Centrifuge 5920 R features an advanced operation system that comes with an easy to use interface to make the operation intuitive and more enjoyable. The multilingual operation menu includes 5 program keys for easy access to your routine programs. For ergonomic lid operation Eppendorf designed the
soft-touch lid closure system. There is no need to exert unnecessary force when closing the centrifuge lid. In combination with a low opening height you can close the centrifuge with ease.

Centrifuge 5920 R features an advanced operation system that comes with an easy to use interface to make the operation intuitive and more enjoyable. The multilingual operation menu includes 5 program keys for easy access
to your routine programs.
-
-
-
-

Application

Centrifugation

Features and Benefits

Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL
Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
High centrifugation speed of up to 21,194 × g (13,700 rpm)
Aerosol-tight Eppendorf QuickLock caps and lids allow fast and ergonomic lid locking
Centrifuge lid with soft-touch lid closure
Quiet operation to improve your work environment
Compact footprint saves valuable bench space
Automatic rotor recognition and imbalance detection for maximum operational safety

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
VisioNize is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service