Key Documents
EP5825719000
Adapter for Eppendorf® A-4-81 MTP/Flex
capacity, 1 x cell culture flask, pack of 2
About This Item
UNSPSC Code:
41103911
NACRES:
NB.78
Pricing and availability is not currently available.
feature
capacity 1 x cell culture flask
Holds 25 cm2 flasks
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5825719000 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with MTP/Flex bucket
General description
Adapter to hold one cell culture flask in Eppendorf rotor A-4-81.
Features and Benefits
  • Adapter to spin down cell cultures directly in a cell culture flask
  • Suitable for 25 cm2 and selective 75 cm2 cell culture flasks
  • Adapters can be used in Eppendorf plate rotor A-4-81-MTP/Flex
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)