Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5820771003

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

F-35-48-17, to hold, 24 x 15 mL conical tubes, with 24 steel sleeves and adapters

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,660.00

$1,660.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$1,660.00

About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

$1,660.00


Please contact Customer Service for Availability

feature

to hold 24 x 15 mL conical tubes
high-speed fixed angle rotor
with 24 steel sleeves and adapters

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5820771003 (Global)

parameter

5500 rpm speed

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

General description

A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service