Key Documents
EP5702747009
Eppendorf® Fixed-angle rotor
F-45-18-17 Cryo, without lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5702747009 is not currently sold in your country. Contact Technical Service
feature
to hold 18 x tubes with screw cap (max o.d. 17 mm)
F-45-18-17 Cryo, without lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5702747009
parameter
4,400 max. RCF (1,970 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5702/5702R/5702RH
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)