Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5495401008

Eppendorf® Rotor lid for rotor FA-24x2

used with centrifuge 5420

Synonym(s):

Aerosol-Tight Rotor Lid for Centrifuge 5420, Aluminum Rotor Lid, Rotor cap

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$500.00

$500.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$500.00

About This Item

UNSPSC Code:
41103913
NACRES:
NB.15

$500.00


Please contact Customer Service for Availability

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5495401008

compatibility

Rotor FA-24x2
used with centrifuge 5420

Looking for similar products? Visit Product Comparison Guide

General description

Rotor lid, for rotor FA-24x2, incl. 1 seal, Centrifuge 5420

Application

CENTRIFUGATION

Features and Benefits

aerosol-tight, aluminumfor Centrifuge 5420

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service