Key Documents
EP5490039009
Rotor for Eppendorf® Concentrator Plus
F-35-8-24, non aerosol tight, to hold, 8 x 25 mL flat-bttom tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5490039009 is not currently sold in your country. Contact Technical Service
feature
to hold 8 x 24 x 86 -90 mm
to hold 8 x 25 mL flat-bttom tubes
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490039009 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
General description
Selection of rotors for the Eppendorf Concentrator Plus.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)