Key Documents
EP5424700004
Eppendorf® Rotor FA-45-24-11-Special
incl. lid, aerosol tight, coated
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5424700004 is not currently sold in your country. Contact Technical Service
feature
to hold 24 x 1.5/2.0 ml tubes
incl. lid, aerosol tight, coated
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5424700004
parameter
15,000 max. RCF (21,130 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5424/5424R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)