Key Documents
EP5416301001
Rotor Key For Eppendorf® Rotors
standard
Synonym(s):
Rotor Key For Eppendorf® Rotors
About This Item
UNSPSC Code:
41103900
NACRES:
NB.82
Product EP5416301001 is not currently sold in your country. Contact Technical Service
feature
standard
manufacturer/tradename
Eppendorf® 5416301001 (Europe)
General description
Centrifuge rotor key for Eppendorf models 5418, 5424 and 5430.
Keys for Eppendorf centrifuge rotors.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)