Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5409711008

Lid for Eppendorf® FA-45-48-11 Rotor, aerosol-tight

aerosol-tight

Synonym(s):

Lid for Eppendorf(R) FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$645.00

$645.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$645.00

About This Item

UNSPSC Code:
41103913
NACRES:
NB.82

$645.00


Please contact Customer Service for Availability

feature

aerosol-tight

manufacturer/tradename

Eppendorf® 5409711008

technique(s)

centrifugation: suitable

compatibility

for use with 5427 R
for use with 5427

Looking for similar products? Visit Product Comparison Guide

General description

  • Replacement rotor lids
  • Aerosol-tight and aluminum lids available

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service