EP5406000160
Eppendorf® Centrifuge 5425 R
keypad control, with rotor FA-24x2, AC/DC input 230 V (GB Plug)
Synonym(s):
Centrifuge
About This Item
Recommended Products
feature
refrigerated
to hold 24 X 2 Ml TUBES
keypad control, with rotor FA-24x2
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5406000160
parameter
-10°-40 ° °C temp. range
15,060 rpm speed
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Outstanding software features (e.g. adjustable acceleration and braking ramps, 3 program buttons (only Centrifuge 5425), short function without continuous pressing of the key)
Extended rotor program (e.g. 5.0 mL rotor, swing-bucket rotor for PCR strips)
OptiBowl® design for quiet operation, even without a rotor lid
Soft-touch one-finger lid closure for ergonomic operation
Max. rotor capacity: 10 × 5.0 mL vessels
Max. speed: 21,300 × g (15,060 rpm)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service