Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP4987000029

Eppendorf® Multipette® E3/E3x/M4

E3x single-channel, with charging cable and Combitips advanced assortment pack (1 Combitip of each size), 1 ul - 50 ml, AC/DC input 100 V / 240 V, 50-60 Hz (EU)

Synonym(s):

Adjustable volume pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01
Product EP4987000029 is not currently sold in your country. Contact Technical Service

feature

CE compliant
E3x single-channel
with charging cable and Combitips advanced assortment pack (1 Combitip of each size)

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 4987000029 (EU)

measuring range

1 ul - 50 ml

volume

1 μL × 50 mL

battery power

rechargeable

Looking for similar products? Visit Product Comparison Guide

General description

  • The expert for long series pipetting and liquids with demanding properties.
  • Dispensing without compromise on accuracy and precision! With the electronic Multipette E3/E3x you save time and money, work stress-free, and get reliable results.
  • It is the ideal and smart solution for filling plates or large series of tubes.
  • The light weight and minimal operation forces of the Multipette E3 reduce the risk of repetitive strain injuries.
  • The Multipette/Combitips® system builds the perfect harmony allowing automatic tip recognition for ease of use. The syringe style Combitips Tip maximizes pipetting accuracy and precision.
  • It is not affected by liquid properties, most manual pipetting errors and provides protection from aerosol contamination.

The Multipette E3/E3x in combination with the Combitips® advanced creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results. The motor driven dispenser gives you full volume control, improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user; filling the tip once allows dispensing up to 100 times. The system is an essential lab tool ideal for all liquid handling applications, accommodating more than 5,000 different dispensing volumes between 1 μL to 50 mL.

Application

  • Repetitive dispensing of volumes in long series such as tube or plate filling, aliquoting reagents and kit usage.
  • Contamination-free dispensing for toxic, radioactive and other hazardous liquids.
  • Precise and accurate dispensing of viscous solutions or liquids with high vapor pressure.
  • Pooling of supernatants.
  • Creation of dilution series Concentration determination.
  • Pooling and volume determination with subsequent dispensing.

Features and Benefits

  • Automatic Combitips recognition: Eliminates time-consuming volume calculations, avoiding incorrect dispensing volumes, Ergonomic one-button tip ejector: For one-handed operation and contact-free Combitips advanced ejection.
  • Speed adjustment: Program aspiration and dispensing speed for the utmost in precision and accuracy while also preventing splashing.
  • Fill the tip once and dispense up to 100 times without a refill.
  • 1 μL to 50 mL dispensing range providing up to 5,000 dispensing volumes with increments as low as 100 nanoliters.
  • Motor-driven function reduces the human error to a minimum and reduces risk of repetitive strain injuries (RSI).
  • Safe and ergonomic operation with 9 different selectable languages.
  • Positive displacement system: Not affected by liquid properties, most manual pipetting errors and provides protection from aerosol contamination.
  • Eppendorf TrackIT compatible: Editable data chip contains all relevant information.

Legal Information

Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service