Documents
EP4960833002
Eppendorf® Bottle Thread Adapter
from GL 32 to GL 40, ETFE
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
material
ETFE
feature
from GL 32 to GL 40
manufacturer/tradename
4960833002
Eppendorf®
Legal Information
Eppendorf is a registered trademark of Eppendorf AG