EP3880000160
Eppendorf® IsoPack and IsoRack
Holds 0.5 ml tubes, parameter (0 °C)
About This Item
Recommended Products
feature
Holds 0.5 ml tubes
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 3880000160
parameter
(0 °C)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service