Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP3123000012

Eppendorf Research® Plus G

single-channel, variable, volume (0.1-2.5 μL), dark gray, includes epT.I.P.S.® box

Synonym(s):

Adjustable volume pipette

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$685.00

$685.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$685.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$685.00


Please contact Customer Service for Availability

feature

includes epT.I.P.S.® box
single-channel, variable

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 3123000012

volume

(0.1-2.5 μL)
-2.5 μL

color

dark gray

Looking for similar products? Visit Product Comparison Guide

General description

The Eppendorf Research plus mechanical pipette combines over 50 years of innovation in liquid handling and is one of the most commonly used pipettes in the world. It provides you with one of the safest and most ergonomic pipettes available today. The Research plus microliter pipette protects the health of our customers during their daily work in the laboratory. The Research plus is the pipette family with the lowest weight and lowest operation forces in the Eppendorf product families. Lower operation forces can only be reached with our Eppendorf electronic pipettes. Enjoy excellent flexibility and choose among single-channel pipettes in fixed or variable volume as well as multi-channel pipette options with 8, 12, 16 and 24 channels.

Product features

It is a good feeling to know you are doing the right thing – to know you are working with one of the most advanced pipettes in the world. A spring loaded tip cone, a secondary calibration option, an improved volume display – and all that in an ultra light, fully autoclavable pipette: That is the Eppendorf Research plus pipette.

Accoladed product

The Research plus mechanical pipette was accoladed with the Bronze “Seal of Quality”. These seals are given to the top 0.1 % of products used in laboratories that have continually received the highest user review ratings. The award is praised as providing “invaluable” insights for scientists and intends to help them choose the ideal laboratory product for their working environment.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service