Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP3116000023

Eppendorf® Pipette Holder System

for 6 Eppendorf Xplorer® or Xplorer® plus, AC/DC input 100 V / 240 V, 50/60 Hz, includes four interchangeable power plugs

Synonym(s):

Eppendorf Pipette Holder, Pipette Holder Stand

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,250.00

$1,250.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$1,250.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$1,250.00


Please contact Customer Service for Availability

feature

Holds 6 x pipetors
includes four interchangeable power plugs

manufacturer/tradename

Eppendorf® 3116000023 (Global)

Looking for similar products? Visit Product Comparison Guide

General description

Keep all your Eppendorf pipettes within good reach and save precious bench space. The new Eppendorf Pipette Holder System provides highly flexible carousels, stands and wall mount devices for all current Eppendorf liquid handling instruments and most predecessors.
The Eppendorf pipette holder system provides safe and convenient storage. As all pipettes are positioned upright, their nose cone does not touch the lab bench reducing the risk of damage and cross contamination. The carousels provide easy access to the pipette of your choice and have a robust design with a small footprint. They can hold up to six liquid handling instruments. The stands carry one electronic pipette or Multipette. Carousels and stands are available with and without charging functionality. The wall mount devices store one manual or electronic pipette or Multipette.

Features and Benefits

New Eppendorf Pipette Holder System stores all current manual and electronic Eppendorf pipettes (including the Multipette) and most of their predecessors
Charger carousel with intelligent charging electronics and increased capacity for up to six instruments
Rotatable holder with completely new design for all manual pipettes including Eppendorf Research®, Eppendorf Research® plus, Reference®, Reference® 2 and Biomaster®
Carousels and stands with interchangeable pipette holders and charger shells, no tools required
Large rubber feet protect carousels and stands from liquids spilled on bench top
Pictograms on all charger shells and pipette holders for clear assignment in the lab

Legal Information

Biomaster is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Xplorer is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service