Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP022363395

Eppendorf® Safe-Lock micro test tubes

capacity 2.0 mL, blue, pkg of 500 ea

Synonym(s):

eppendorfs

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$121.00

$121.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$121.00

About This Item

UNSPSC Code:
41121703

$121.00


Please contact Customer Service for Availability

material

blue
polypropylene (push fit
)

round bottom

sterility

non-sterile

feature

RCF 30,000  × g

packaging

pkg of 500 ea

manufacturer/tradename

Eppendorf® 022363395

capacity

2.0 mL

diam.

10.5 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Eppendorf Safe-Lock Tubes, 2.0 mL, Eppendorf Quality, blue, 500 tubes
  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • g-Safe®: Depending on Safe-Lock Tube variant (see manual) exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Manufactured without mold release agents, softeners or biocides – Materials which could have a negative effect on bioassays
  • g-Safe®: Exceptional centrifugation stability up to 25,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • Also available in purity grades of lot-certified PCR clean, Biopur® and Eppendorf Forensic DNA Grade
  • Also available in variants Eppendorf Protein LoBind or Eppendorf DNA LoBind for maximum recovery of valuable samples and in amber for light-sensitive samples

Features and Benefits

Eppendorf Safe-Lock Tubes offer special protection against unintentional opening and completely eminate any danger of leaks during incubation and storage

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service