Skip to Content
MilliporeSigma
All Photos(1)

Documents

EP0030132947

Eppendorf® Masterclear® Real-time PCR Film

PCR clean, self-adhesive, pkg of 100 ea

Synonym(s):

PCR, Plates, Sealing films

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.35

sterility

non-sterile

feature

PCR clean
self-adhesive

packaging

pkg of 100 ea

manufacturer/tradename

Eppendorf® 0030132947

technique(s)

PCR: suitable

Application

Eppendorf® Masterclear® Real-time PCR Film is suitable for effective and tight sealing of polymerase chain reation (PCR) plates.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Masterclear is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service