Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030129547

EPPENDORF TWIN.TEC® PCR PLATES 384

PCR Clean, colorless, pkg of 25 ×

Synonym(s):

Eppendorf twin.tec® PCR Plates 384, 96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$443.00

$443.00


Please contact Customer Service for Availability


Select a Size

Change View
25 EA
$443.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.15
material:
colorless
colorless wells
size:
384 well
sterility:
non-sterile
binding type:
low binding surface
feature:
skirt
PCR Clean

$443.00


Please contact Customer Service for Availability

material

colorless
colorless wells

sterility

non-sterile

feature

skirt
PCR Clean

manufacturer/tradename

Eppendorf® 0030129547 (Global)

packaging

pkg of 25 ×

technique(s)

PCR: suitable

size

384 well

wells

384 wells

working volume

40 μL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
twin.tec PCR Plates LoBind improve recovery of nucleic acids by reducing their absorption to the tube wall. A combination of special manufacturing technologies and selected polypropylene batches ensures nearly 100 % recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination. twin.tec PCR Plates LoBind are batch-tested and certified by an independent laboratory to be free from DNA, DNase, RNase and PCR inhibitors.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service