EP0030129520
Eppendorf® twin.tec(R) PCR plate 96 LoBind
semi-skirted, with barcode(s), clear, set of 25 pieces
About This Item
semi-skirted, with barcode(s)
Recommended Products
feature
skirt
semi-skirted, with barcode(s)
packaging
set of 25 pieces
manufacturer/tradename
Eppendorf® 0030129520
technique(s)
PCR: suitable
size
96 well
color
clear
binding type
low binding surface
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service