EP0030078560
Eppendorf® ep Dualfilter T.I.P.S.®
PCR clean, sterile, tip volume × L 20-300 μL × 55 mm, orange tip, case of 960 tips (10 racks x 96 tips)
Synonym(s):
Dualfilter tips, Filter tips
Select a Size
$256.00
Select a Size
About This Item
sterile
PCR clean
RNase and DNase free
filter low retention etc.
$256.00
Recommended Products
sterility
sterile (PCR clean/sterile)
sterile
feature
DNA free
PCR clean
RNase and DNase free
filter low retention etc.
packaging
case of 960 tips (10 racks x 96 tips)
manufacturer/tradename
Eppendorf® 0030078560
tip volume × L
20-300 μL × 55 mm
volume
20—300 μL
color
orange tip
binding type
low binding surface
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
The Eppendorf ep Dualfilter T.I.P.S.® are the first filtertips with a two phase filter for contamination protection. The tips are manufactured to the highest possible quality from pure, non recycled materials under cleanroom conditions. They are characterized by defined flow dynamics, low wettability and high thermal stability. They are certified pyrogen free and PCR clean (free from human DNA, DNase, RNase and PCR-inhibiting substances).
These ep Dualfilter T.I.P.S.® contain well-defined different pore sizes within the two filter layers. The white layer which faces the sample retains drops, splashes and aerosols. The blue layer above it forms a highly-effective double barrier, that reliably binds even biomolecules.
Without any kind of filter the pipette is exposed to contamination by sample and aerosols. Conventional single layer filters do not fully block particles and molecules. Only ep Dualfilter T.I.P.S.® provide reliable protection against even the finest impurities.
Features and Benefits
- Two filter layers provide dual protection for pipette and sample
- Two filter layers provide dual protection from aerosols and biomolocules
- Free from PCR inhibitor additives
- Patent pending technology
- All tips are provided sterile, Eppendorf PCR clean and pyrogen-free
- Batch certified
- Defined flow dynamics
- Low wettability
- High thermal stability
- Leak proof seal
- Minimum attachment and ejection forces
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service