Documents
EP0030073410
Eppendorf® epT.I.P.S.® G Reloads, Eppendorf Quality
volume (0.5-20 μL), L (46 mm), light gray, pack of 960 ea (10 trays × 96 tips)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
material
gray (Tray)
sterility
non-sterile
packaging
pack of 960 ea (10 trays × 96 tips)
manufacturer/tradename
Eppendorf® 0030073410
L
(46 mm)
maximum volume
20 μL
volume
(0.5-20 μL)
color
light gray
General description
The pipette tips are single-use pipette tips for aspirating and dispensing liquids with air-cushion pipettors. The packaging of the pipette tips is color-coded and corresponds to the color of the control button on the Eppendorf pipettor.
Features and Benefits
  • Single-use pipette tips
  • Packaged as reloade
  • Universal cone
  • Easy handling
  • Avaiable in different quality grades
  • Minimal force required for attachment or ejection
  • Different volume sizes with color coding
  • Volume marking for visual inspection
  • Seal completely
  • Provide maximum precision and accuracy
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Quality is a trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG