EP0030000838
Eppendorf® epT.I.P.S.® standard
volume range 0.1-20 μL, pack of 1000 ea (2 bags × 500 tips)
Synonym(s):
pipetting tips
About This Item
Recommended Products
material
gray
polypropylene
sterility
non-sterile
packaging
pack of 1000 ea (2 bags × 500 tips)
manufacturer/tradename
Eppendorf® 0030000838
maximum volume
20 μL
volume range
0.1-20 μL
volume
0.1—20 μL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- High-quality and original pipettes packed in resealable bags
- Tips are autoclave-friendly at 121°C for 20 mins
- The fine retracted or extended tips enable simpler ways to work with small sample volumes
- Tips are universal that can be used with Eppendorf pipettes and pipettes from other manufacturers
- Disposable racks with a slim design, reclosable and easy stackability
- The cone geometry of the tips ensures optimal pipette fit with Eppendorf pipettes Fine graduations make a visual examination of the pipetting volume even easier
- Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service